Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGR2 Recombinant Rabbit Monoclonal Antibody (JG78-39)
GREENER_CHOICE

Catalog No. PIMA534770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534770 Supplier Invitrogen™ Supplier No. MA534770
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: MCF-7, SH-SY-5Y, mouse kidney tissue.

The EGR2 gene provides instructions for making a protein called early growth response 2, which is part of the early growth response family of proteins. These proteins bind to specific areas of DNA and help control the activity of particular genes. On the basis of this action, the proteins are referred to as transcription factors. The early growth response 2 protein activates several genes that are involved in the formation and maintenance of myelin, the fatty substance that covers and protects nerve cells. Myelin promotes the efficient transmission of nerve impulses. If myelin is lost (demyelination) or its structure is disrupted, the transmission of nerve impulses is impaired. Mutations in the EGR2 gene can cause two forms of Charcot-Marie-Tooth disease, type 1D or type 4E (sometimes called congenital hypomyelinating neuropathy) or a severe form of type 1D (sometimes called Dejerine-Sottas syndrome) that begins during infancy or early childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGR2
Applications Western Blot
Classification Recombinant Monoclonal
Clone JG78-39
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Egr2
Gene Accession No. P11161
Gene Alias AT591; CMT1D; CMT4E; DKFZp686J1957; E3 SUMO-protein ligase EGR2; E3 SUMO-protein transferase ERG2; early growth response 2; early growth response protein 2; Egr2; Egr-2; FLJ1454; FLJ14547; Krox20; Krox-20; Krox-20 homolog; KROX-20, Drosophila, homolog (early growth response-2); NGF1-B; OTTHUMP00000062133; Zfp-25; Zfp-6; Zinc finger protein Krox-20
Gene Symbols Egr2
Host Species Rabbit
Immunogen Recombinant protein within Human EGR2 aa 2-96.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1959
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.