Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582238 Supplier Invitrogen™ Supplier No. PA582238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPVSFHDPQG NPGLAYSPQD YQSAKPALDS NLFPMIPDYN LYHHPNDMGS IPEHKPFQGM DPIRVNPPPI TPLETIKAFK DKQIHPGFGS LPQPPLTLKP IRPRKYPNRP SKTPLHERPH ACPAEGCDRR FSRSD Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a transcriptional regulator that belongs to the EGR family of C2H2-type zinc-finger proteins. It is an immediate-early growth response gene which is induced by mitogenic stimulation. The protein encoded by this gene participates in the transcriptional regulation of genes in controling biological rhythm. It may also play a role in a wide variety of processes including muscle development, lymphocyte development, endothelial cell growth and migration, and neuronal development. Alternative splicing results in multiple transcript variants encoding distinct isoforms.[provided by RefSeq, Dec 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGR3
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EGR3
Gene Accession No. Q06889
Gene Alias early growth response 3; early growth response protein 3; EGR3; EGR-3; MGC138484; Pilot; zgc:92020; zinc finger protein pilot
Gene Symbols EGR3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EGR3. Recombinant protein control fragment (Product #RP-89413).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.