Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGR3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5103317

Catalog No. PIPA5103317


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of total EGR3.

This gene encodes a transcriptional regulator that belongs to the EGR family of C2H2-type zinc-finger proteins. It is an immediate-early growth response gene which is induced by mitogenic stimulation. The protein encoded by this gene participates in the transcriptional regulation of genes in controling biological rhythm. It may also play a role in a wide variety of processes including muscle development, lymphocyte development, endothelial cell growth and migration, and neuronal development. Alternative splicing results in multiple transcript variants encoding distinct isoforms.[provided by RefSeq, Dec 2010].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EGR3
Polyclonal
Unconjugated
EGR3
early growth response 3; early growth response protein 3; EGR3; EGR-3; MGC138484; Pilot; zgc:92020; zinc finger protein pilot
Rabbit
Affinity chromatography
RUO
13655, 1960, 25148
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P43300, P43301, Q06889
EGR3
A synthesized peptide derived from human EGR3(Accession Q06889), corresponding to amino acid residues T124-Q174.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.