Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556028 Supplier Invitrogen™ Supplier No. PA556028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PWELLSVGAP GNCGSQGDYQ AAPEARFPVI GTKIEDLLSI SCPAELPAVP ANRLYPSGAY DAFPLAPGDL GEGAEGLPGL LTPPSGEGGS SGDGGEFLAS TQPQLSPLGL RSAAAADFPK PL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.

Early growth response protein 4 (EGR-4), also known as AT133, is a protein that in humans is encoded by the EGR4 gene. EGR-4 is a member of the early growth response (EGF) family of zinc finger transcription factors. Transcriptional regulator. Recognizes and binds to the DNA sequence 5'-GCGGGGGCG-3' (GSG). Activates the transcription of target genes whose products are required for mitogenesis and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGR4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EGR4
Gene Accession No. Q05215
Gene Alias AT133; early growth response 4; early growth response protein 4; Early response protein NGFI-C; Egr4; EGR-4; Egr4l1; Nerve growth factor-induced protein C; NGF1-C; Ngfic; NGFI-C; PAT133; zinc-finger transcription factor NGFI-C (early response gene)
Gene Symbols EGR4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EGR4. Recombinant protein control fragment (Product #RP-100716).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1961
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.