Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595746 Supplier Thermo Scientific Supplier No. PA595746
Add to Cart
Add to Cart

EGR4 Polyclonal Antibody

EGR4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen EGR4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EGR4
Gene Accession No. Q9WUF2
Gene Alias AT133; early growth response 4; early growth response protein 4; Early response protein NGFI-C; Egr4; EGR-4; Egr4l1; Nerve growth factor-induced protein C; NGF1-C; Ngfic; NGFI-C; PAT133; zinc-finger transcription factor NGFI-C (early response gene)
Gene Symbols EGR4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-367 of mouse Egr4 (NP_0656211).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13656
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.