Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EGR4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5141024
This item is not returnable.
View return policy
Description
EGR4 Polyclonal Antibody for IHC (P)
Early growth response protein 4 (EGR-4), also known as AT133, is a protein that in humans is encoded by the EGR4 gene. EGR-4 is a member of the early growth response (EGF) family of zinc finger transcription factors. Transcriptional regulator. Recognizes and binds to the DNA sequence 5'-GCGGGGGCG-3' (GSG). Activates the transcription of target genes whose products are required for mitogenesis and differentiation.
Specifications
EGR4 | |
Polyclonal | |
Unconjugated | |
EGR4 | |
AT133; early growth response 4; early growth response protein 4; Early response protein NGFI-C; Egr4; EGR-4; Egr4l1; Nerve growth factor-induced protein C; NGF1-C; Ngfic; NGFI-C; PAT133; zinc-finger transcription factor NGFI-C (early response gene) | |
Rabbit | |
Protein A | |
RUO | |
13656, 1961, 25129 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q00911, Q05215, Q9WUF2 | |
EGR4 | |
Recombinant protein within mouse EGR4 aa 300-478/589. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction