Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EGTA
GREENER_CHOICESDP

Catalog No. 28071G Shop All Novus Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-071-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 28-071-G Supplier R&D Systems™ Supplier No. 2807/1G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Calcium chelating agent

Calcium chelator; protects against cell death caused by nitric oxide-induced calcium influx into nerve cells.

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10

Specifications

Quantity 1 g
Product Line Tocris
Formula Weight 380.35
Purity >98%
Molecular Weight (g/mol) 380.35
Chemical Name or Material EGTA
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.