Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557281 Supplier Invitrogen™ Supplier No. PA557281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSPTSKLGYS YSRDLDLAKK KHASLRQTES DPDADRTTLN HADHSSKIVQ HRLLSRQEEL KERARVLLEQ ARRDAALKAG NKHNTNTATP F Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 76%.

This gene encodes an Eps15 homology domain binding protein. The encoded protein may play a role in endocytic trafficking. A single nucleotide polymorphism in this gene is associated with an aggressive form of prostate cancer. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ehbp1
Gene Accession No. Q8NDI1
Gene Alias AF424697; DNA sequence AF424697; EH domain binding protein 1; EH domain-binding protein 1; EHBP1; Flj21950; HPC12; Kiaa0903; NACSIN; NPF calponin-like protein; RGD1310604; testis tissue sperm-binding protein Li 50e
Gene Symbols Ehbp1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EHBP1. Recombinant protein control fragment (Product #RP-96020).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23301
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.