Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EHBP1L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140037
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: EVLGTQKTEA GGSGVLQTRT TIAETEVLVT QEISGDLGPL KIEDTIQSEM LGTQETEVEA SRVPESEAEG TEAKILGTQE ITARDSGV.

Specifications
EHBP1L1 | |
Polyclonal | |
Unconjugated | |
EHBP1L1 | |
EH domain binding protein 1 like 1; EH domain binding protein 1-like 1; EH domain-binding protein 1-like protein 1; EHBP1L1; G430002G23Rik; Tangerin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254102 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N3D4 | |
EHBP1L1 | |
Recombinant protein corresponding to Human EHBP1L1. Recombinant protein control fragment (Product #RP-109348). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction