Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561497 Supplier Invitrogen™ Supplier No. PA561497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HDIAKLMPLL RQEELESTEV GVQGGAFEGT HMGPFVERGP DEAMEDGEEG SDDEAEWVVT KDKSKYDEIF YNLAPADGKL SGSKAKTWM Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

This gene encodes a member of the EH domain-containing protein family. These proteins are characterized by a C-terminal EF-hand domain, a nucleotide-binding consensus site at the N terminus and a bipartite nuclear localization signal. The encoded protein interacts with the actin cytoskeleton through an N-terminal domain and also binds to an EH domain-binding protein through the C-terminal EH domain. This interaction appears to connect clathrin-dependent endocytosis to actin, suggesting that this gene product participates in the endocytic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ehd2
Gene Accession No. Q9NZN4
Gene Alias BC027084; C130052H20Rik; EH domain containing 2; EH domain-containing protein 2; Ehd2; EH-domain containing 2; FLJ96617; MGEPS; PAST homolog 2; PAST2; putative eps protein
Gene Symbols Ehd2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EHD2. Recombinant protein control fragment (Product #RP-98852).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30846
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.