Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206850
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6850 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6850 Supplier Fabgennix Inc Supplier No. EHD2201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EHD2 Polyclonal antibody specifically detects EHD2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen EHD2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Ehd2
Gene Accession No. Q4V8H8, Q8BH64, Q9NZN4
Gene Alias BC027084; C130052H20Rik; EH domain containing 2; EH domain-containing protein 2; Ehd2; EH-domain containing 2; FLJ96617; MGEPS; PAST homolog 2; PAST2; putative eps protein
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid 420-450 on human EH-Domain Containing Protein 2.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259300, 30846, 361512
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.