Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EHD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$336.16 - $690.48
Specifications
| Antigen | EHD3 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Immunohistochemistry-Frozen |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
EHD3 Polyclonal specifically detects EHD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| EHD3 | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EH domain containing 3, EH domain-containing protein 3, EHD2, EH-domain containing 3, PAST homolog 3, PAST3 | |
| EHD3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9NZN3 | |
| 30845 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PDNRKLFEAEEQDLFRDIQSLPRNAALRKLNDLIKRARLAKVHAYIISSLKKE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title