Learn More
Invitrogen™ EHD3 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | EHD3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | EHD3 |
| Gene Accession No. | Q9NZN3, Q9QXY6 |
| Gene Alias | EH domain containing 3; EH domain-containing protein 3; Ehd2; Ehd3; EH-domain containing 3; EH-domain containing protein 2; PAST homolog 3; PAST3; si:ch211-147c1.2 |
| Gene Symbols | EHD3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.