Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562079 Supplier Invitrogen™ Supplier No. PA562079
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLYKSKLLPL EEHYRFHEFH SPALEDADFD NKPMVLLVGQ YSTGKTTFIR Y Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The EHD3 gene encodes the EH-domain containing protein 3, part of the EHD protein family involved in the regulation of endocytic recycling and membrane trafficking. EHD3 plays a crucial role in the endosomal transport system, impacting how receptors and other membrane proteins are recycled back to the cell surface after internalization. This protein aids in maintaining cellular homeostasis by ensuring the efficient sorting and transport of cargo through the endosomal system. EHD3 has been implicated in various cellular functions, including insulin signaling, receptor recycling, and neuronal development, and significantly influences synaptic transmission and plasticity. Alterations in EHD3 expression or function can disrupt endosomal trafficking processes and are linked to conditions such as neurodegenerative diseases and cancer. Research on EHD3 is focused on uncovering its role in membrane dynamics and identifying potential therapeutic strategies for disorders involving dysfunctional endocytic pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EHD3
Gene Accession No. Q9NZN3
Gene Alias EH domain containing 3; EH domain-containing protein 3; Ehd2; Ehd3; EH-domain containing 3; EH-domain containing protein 2; PAST homolog 3; PAST3; si:ch211-147c1.2
Gene Symbols EHD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EHD3. Recombinant protein control fragment (Product #RP-100845).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.