Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734857 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4857 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4857 Supplier Proteintech Group Inc Supplier No. 253201AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EHD3 plays a role in endocytic transport.

Specifications

Antigen EHD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene EHD3
Gene Accession No. Q9NZN3, Q9QXY6
Gene Alias EH domain containing 3, EH domain containing protein 3, EHD2, EHD3, PAST homolog 3, PAST3
Gene Symbols EHD3
Host Species Rabbit
Immunogen EHD3 Fusion Protein Ag18216
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30845, 57440
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.