Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117926 Supplier Invitrogen™ Supplier No. PA5117926
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EHD4 Polyclonal Antibody for Western Blot, IP

ATP- and membrane-binding protein that probably controls membrane reorganization/tubulation upon ATP hydrolysis. Plays a role in early endosomal transport. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHD4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene EHD4
Gene Accession No. Q9H223
Gene Alias 2210022F10Rik; AI197390; AI846352; AV006278; EH domain containing 4; EH domain containing protein MPAST2; EH domain-containing protein 4; EHD4; EH-domain containing 4; FKSG7; HCA10; HCA11; Hepatocellular carcinoma-associated protein 10/11; hepatocellular carcinoma-associated protein HCA11; mPAST2; ortholog of rat pincher; PAST homolog 2; PAST homolog 4; Past2; PAST4; pincher; pinocytic chaperone
Gene Symbols EHD4
Host Species Rabbit
Immunogen E. coli-derived Human EHD4 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30844
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.