Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206856
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6856 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6856 Supplier Fabgennix Inc Supplier No. EHD4401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EHD4 Polyclonal antibody specifically detects EHD4 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen EHD4
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene EHD4
Gene Accession No. Q9EQP2, Q9H223
Gene Alias 2210022F10Rik; AI197390; AI846352; AV006278; EH domain containing 4; EH domain containing protein MPAST2; EH domain-containing protein 4; EHD4; EH-domain containing 4; FKSG7; HCA10; HCA11; Hepatocellular carcinoma-associated protein 10/11; hepatocellular carcinoma-associated protein HCA11; mPAST2; ortholog of rat pincher; PAST homolog 2; PAST homolog 4; Past2; PAST4; pincher; pinocytic chaperone
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 420-450 on EH-domain containing 4 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192204, 30844, 98878
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.