Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563890
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563890 Supplier Invitrogen™ Supplier No. PA563890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MILEGGGVMN LNPGNNLLHQ PPAWTDSYST CNVSSGFFGG QWHEIHPQYW TKYQVWEWLQ HLLDTNQLDA NCIPFQEFDI NGEHL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.

This gene encodes a protein that belongs to an ETS transcription factor subfamily characterized by epithelial-specific expression (ESEs). The encoded protein acts as a transcriptional repressor and may be associated with asthma susceptibility. This protein may be involved in epithelial differentiation and carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EHF
Gene Accession No. Q9NZC4
Gene Alias 9030625L19Rik; AU019492; EHF; Epithelium-specific Ets transcription factor 3; ESE3; ESE-3; ESE3 transcription factor; ESE3B; ESEJ; ETS domain-containing transcription factor; ETS homologous factor; hEHF; Unknown (protein for MGC:133971)
Gene Symbols EHF
Host Species Rabbit
Immunogen Recombinant protein fragment of Human EHF (Product #RP-103380)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26298
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.