Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHHADH Mouse anti-Human, Clone: 7F6AF11, Abnova™

Catalog No. 89035438 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-438 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-438 Supplier Abnova Corporation Supplier No. MAB14033

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human EHHADH.

  • High specificity
  • High reactivity

Specifications

Antigen EHHADH
Applications Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 7F6AF11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human EHHADH.
Dilution Immunocytochemistry (1 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. Q08426
Gene Alias ECHD/LBFP/LBP/MGC120586/PBFE
Gene Symbols EHHADH
Host Species Mouse
Immunogen Human liver mitochondria.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1962
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.