Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHMT1 Monoclonal Antibody (1B7B4)
GREENER_CHOICE

Catalog No. PIMA515277
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515277 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515277 Supplier Invitrogen™ Supplier No. MA515277
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15277 targets GLP in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15277 immunogen is purified recombinant fragment of GLP expressed in E. Coli.

EHMT1, also known as EUHMTASE1, is a histone methyltransferase. This protein methylates 'Lys-9' of histone H3 in vitro. H3 'Lys-9' methylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 proteins to methylated histones. EHMT1 is Probably targeted to histone H3 by different DNA-binding proteins like E2F6, MGA, MAX and/or DP1. During G0 phase, it probably contributes to silencing of MYC- and E2F-responsive genes, suggesting a role in the G0/G1 transition of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHMT1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B7B4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene EHMT1
Gene Accession No. Q9H9B1
Gene Alias 9230102N17Rik; bA188C12.1; D330003E03; EHMT1; euchromatic histone lysine methyltransferase 1; euchromatic histone methyltransferase 1; euchromatic histone-lysine N-methyltransferase 1; Euhmtase1; Eu-HMTase1; FP13812; G9a-like protein 1; GLP; GLP1; H3-K9-HMTase 5; histone H3-K9 methyltransferase 5; Histone-lysine N-methyltransferase EHMT1; histone-lysine N-methyltransferase, H3 lysine-9 specific 5; KIAA1876; KMT1D; lysine N-methyltransferase 1D; mKIAA1876
Gene Symbols EHMT1
Host Species Mouse
Immunogen Purified recombinant fragment of GLP expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79813
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.