Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHMT2 (G9a) Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
SDP

Mouse Recombinant Monoclonal Antibody

$110.70 - $492.00

Specifications

Antigen EHMT2 (G9a)
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3760
SDP
Encompass
View Documents
Active Motif
91230
10 μg
Each for $110.70
Only null left
Add to Cart
 
50-199-3759
SDP
Encompass
View Documents
Active Motif
91229
100 μg
Each for $492.00
Only null left
Add to Cart
 
Description

Description

A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. This gene is found near this cluster; it was mapped near the gene for C2 within a 120-kb region that included a HSP70 gene pair. These genes are all within the human major histocompatibility complex class III region. This gene was thought to be two different genes, NG36 and G9a, adjacent to each other but a recent publication shows that there is only a single gene. The protein encoded by this gene is thought to be involved in intracellular protein-protein interaction. There are three alternatively spliced transcript variants of this gene but only two are fully described.
Specifications

Specifications

EHMT2 (G9a)
Western Blot
Unconjugated
Mouse
Human
Q96KQ7
10919
This antibody was raised against full length human recombinant EHMT2 (G9a) protein.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Ehmt2
1 mg/mL
Monoclonal
Liquid
RUO
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5
ankyrin repeat-containing protein; Bat8; BOS_21678; C6orf30; D17Ertd710e; DKFZp686H08213; EHMT2; euchromatic histone lysine methyltransferase 2; euchromatic histone lysine N-methyltransferase 2; Euchromatic histone-lysine N-methyltransferase 2; FLJ35547; G9a; G9A histone methyltransferase; GAT8; H3 ly; H3-K9-Histone-Methyl-Transferase; H3-K9-HMTase 3; Histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B associated transcript 8, rat orthologue; HLA-B-associated transcript 8; KMT1C; Lysine N-methyltransferase 1C; Ng36; NG36/G9a; Protein G9a; SBAB-707F1.1
Ehmt2
IgG2a
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.