Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHMT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588568 Supplier Invitrogen™ Supplier No. PA588568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGNRAIRTEK IICRDVARGY ENVPIPCVNG VDGEPCPEDY KYISENCETS TMNIDRNITH LQHCTCVDDC SSSNCLCGQL SIRCWYDKDG RLLQEFNKIE PPLIFECNQA CSCWRNCKNR VVQSGIKVRL QLYRTAKMGW GVRALQTIPQ GTFICEYVGE LISDAEADVR EDDSYLFDLD NKDGEVYCID ARYYGNISRF INHLCDPNII PVRVFMLHQD LRFPRIAFFS SRDIRTGEEL GFDYGDRFWD IKSKYFTCQC GSEKCKHSAE AIALEQSRLA RLDPHPELLP ELGSLPPVNT; Positive Samples: U-937, 293T, Jurkat, MCF7, HepG2; Cellular Location: Chromosome, Nucleus.

A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. This gene is found near this cluster; it was mapped near the gene for C2 within a 120-kb region that included a HSP70 gene pair. These genes are all within the human major histocompatibility complex class III region. This gene was thought to be two different genes, NG36 and G9a, adjacent to each other but a recent publication shows that there is only a single gene. The protein encoded by this gene is thought to be involved in intracellular protein-protein interaction. There are three alternatively spliced transcript variants of this gene but only two are fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHMT2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ehmt2
Gene Accession No. Q96KQ7, Q9Z148
Gene Alias ankyrin repeat-containing protein; Bat8; C6orf30; D17Ertd710e; DKFZp686H08213; Ehmt2; euchromatic histone lysine methyltransferase 2; euchromatic histone lysine N-methyltransferase 2; euchromatic histone-lysine N-methyltransferase 2; FLJ35547; G9a; G9A histone methyltransferase; GAT8; H3 ly; H3-K9-Histone-Methyl-Transferase; H3-K9-HMTase 3; histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B associated transcript 8, rat orthologue; HLA-B-associated transcript 8; KMT1C; lysine N-methyltransferase 1C; NG36; NG36/G9a; Protein G9a
Gene Symbols Ehmt2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 877-1176 of human EHMT2 (NP_0795325).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10919, 110147, 361798
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.