Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EHT 1864
GREENER_CHOICESDP

Catalog No. 387250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-725-0 50 mg
38-721-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-725-0 Supplier R&D Systems™ Supplier No. 3872/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent Rac family inhibitor

Inhibitor of Rac family GTPases. Blocks activation by direct binding to Rac1, Rac1b, Rac2 and Rac3 (KD values are 40, 50, 60 and 250 nM respectively). Inhibits Rac, Ras and Tiam-induced growth transformation of NIH-3T3 fibroblasts. Reduces β-amyloid peptide production in vivo.

Chemical Identifiers

CAS 754240-09-0
Molecular Formula C25H29Cl2F3N2O4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 581.47
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 581.472
Chemical Name or Material Rho GTPase Inhibitors
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.