Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EI24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551262
Description
The antibody detects endogenous levels of total EI24 protein.
This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.
Specifications
EI24 | |
Polyclonal | |
Unconjugated | |
EI24 | |
AA536736; AI115355; ectopic P-granules autophagy protein 4 homolog; EI24; EI24, autophagy associated transmembrane protein; EPG4; etoposide induced 2.4; etoposide induced 2.4 mRNA; etoposide-induced protein 2.4; Etoposide-induced protein 2.4 homolog; p53-induced gene 8 protein; Pig8; TP53I8; tumor protein p53 inducible protein 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9538 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O14681 | |
EI24 | |
Synthetic peptide of human EI24. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction