Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EI24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562296
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562296 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562296 Supplier Invitrogen™ Supplier No. PA562296
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HKTVYLQSAL SSSTSAEKFP SPHPSPAKLK A Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EI24
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EI24
Gene Accession No. O14681
Gene Alias AA536736; AI115355; ectopic P-granules autophagy protein 4 homolog; EI24; EI24, autophagy associated transmembrane protein; EPG4; etoposide induced 2.4; etoposide induced 2.4 mRNA; etoposide-induced protein 2.4; Etoposide-induced protein 2.4 homolog; p53-induced gene 8 protein; Pig8; TP53I8; tumor protein p53 inducible protein 8
Gene Symbols EI24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EI24. Recombinant protein control fragment (Product #RP-104070).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9538
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.