Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EI24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572705
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572705 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572705 Supplier Invitrogen™ Supplier No. PA572705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EI24 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EI24
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene EI24
Gene Accession No. O14681, Q4KM77, Q61070
Gene Alias AA536736; AI115355; ectopic P-granules autophagy protein 4 homolog; EI24; EI24, autophagy associated transmembrane protein; EPG4; etoposide induced 2.4; etoposide induced 2.4 mRNA; etoposide-induced protein 2.4; Etoposide-induced protein 2.4 homolog; p53-induced gene 8 protein; Pig8; TP53I8; tumor protein p53 inducible protein 8
Gene Symbols EI24
Host Species Rabbit
Immunogen EI24 antibody for a 19 amino acid synthetic peptide near the carboxy terminus of human EI24. The immunogen is within the last 50 amino acids of EI24.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13663, 300514, 9538
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.