Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EI24 Rabbit anti-Human, Mouse, Rat, Bovine, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260709
Description
EI24 Polyclonal specifically detects EI24 in Human, Mouse, Rat, Bovine samples. It is validated for Immunoprecipitation.Specifications
EI24 | |
Polyclonal | |
Immunoprecipitation 8-15 ul/mg lysate | |
EPG4, etoposide induced 2.4 mRNA, p53-induced gene 8 protein, PIG8etoposide-induced protein 2.4 homolog, TP53I8, tumor protein p53 inducible protein 8 | |
Rabbit | |
Affinity Purified | |
Apoptosis | |
9538 | |
Store at 4degreesC. |
Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
EI24 | |
between 290 and 340 | |
100 μL | |
Primary | |
Human, Mouse, Rat, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction