Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EI2BL Antibody (OTI9H7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272280H
Description
EI2BL Monoclonal specifically detects EI2BL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EI2BL | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MRI1 | |
Full length recombinant protein of human EI2BL (NP_115661) produced in HEK293T cells. | |
0.1 mL | |
84245.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI9H7 | |
Western Blot, Immunohistochemistry | |
EC 5.3.1.23, M1Pi, Mediator of RhoA-dependent invasion, methylthioribose-1-phosphate isomerase, methylthioribose-1-phosphate isomerase homolog (S. cerevisiae), MGC3207, MRDI, MTNA, MTR-1-P isomerase, S-methyl-5-thioribose-1-phosphate isomerase, Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein, Ypr118w | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction