Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium borohydride, 95%
Click to view available options
Quantity:
5 g
10 g
50 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Melting Point | 280.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 10 g |
| Assay Percent Range | 95% |
| Linear Formula | LiBH4 |
| Fieser | 01,603; 04,296; 11,293; 12,276; 13,92; 14,191; 15,186 |
| Merck Index | 15, 5581 |
| Solubility Information | Solubility in water: 209g/L (10°C). Other solubilities: soluble in ether, ethanol, tetrahydrofuran and, aliphatic amines |
| IUPAC Name | lithium;boron(1-) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Reacts violentl |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face p |
| EINECSNumber | 241-021-7 |
| RTECSNumber | ED2725000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction