Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EID1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115165 Supplier Invitrogen™ Supplier No. PA5115165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EID1.

E1A-like inhibitor of differentiation-1 (EID-1), an acetyltransferase enzyme, binds both the retinoblastoma protein (Rb), a regulator of cell cycle and tissue specific transcription, and the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. EID-1 inhibits cellular differentiation by blocking the histone acetyltransferase activity of p300. EID-1 also acetylates both histones and non-histone proteins such as NCOA3 co-activator. By acetylating histones, EID-1 gives a specific tag for transcriptional activation. In addition to binding Rb and p300, EID-1 also binds to phosphorylated CREB protein, mediating cAMP gene regulation. EID-1 augments the activity of phosphorylated CREB and activates transcription of cAMP responsive genes as a co-activator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EID1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene EID1
Gene Accession No. Q9Y6B2
Gene Alias 21 kDa pRb-associated protein; 2610002K20Rik; C15orf3; CREBBP/EP300 inhibitor 1; CREBBP/EP300 inhibitory protein 1; CRI1; E1A-like inhibitor of differentiation 1; EID; eid1; EID-1; EP300 interacting inhibitor of differentiation 1; EP300-interacting inhibitor of differentiation 1; IRO45620; NB4 apoptosis related protein; ORF12; PNAS-22; PTD014; Rb- and p300-binding protein EID-1; RBP21; retinoblastoma protein-associated protein; RGD1562702
Gene Symbols EID1
Host Species Rabbit
Immunogen A synthesized peptide derived from human EID1(Accession Q9Y6B2), corresponding to amino acid residues K72-Y122.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23741
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.