Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EID2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561012
Description
Immunogen sequence: AREAAFDAEY QRNPHRVDLD ILTFTIALTA SEVINPLIEE LGCDKFINRE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
EID2 interacts with EP300 and acts as a repressor of MYOD-dependent transcription and muscle differentiation. Inhibits EP300 histone acetyltransferase activity. Acts as a repressor of TGFB/SMAD transcriptional responses. It may act as a repressor of the TGFB/SMAD3-dependent signaling by selectively blocking formation of TGFB-induced SMAD3-SMAD4 complex.
Specifications
EID2 | |
Polyclonal | |
Unconjugated | |
EID2 | |
CREBBP/EP300 inhibitor 2; CREBBP/EP300 inhibitory protein 2; Cri2; E1A like inhibitor of differentiation 2; EID-1-like inhibitor of differentiation 2; Eid2; EID-2; EP300 interacting inhibitor of differentiation 2; EP300-interacting inhibitor of differentiation 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
163126 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6I1 | |
EID2 | |
Recombinant protein corresponding to Human EID2. Recombinant protein control fragment (Product #RP-99182). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction