Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EID2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA525543

Catalog No. PIPA525543


Only null left
Add to Cart

Description

Description

EID2 Polyclonal Antibody for Western Blot

EID2 interacts with EP300 and acts as a repressor of MYOD-dependent transcription and muscle differentiation. Inhibits EP300 histone acetyltransferase activity. Acts as a repressor of TGFB/SMAD transcriptional responses. It may act as a repressor of the TGFB/SMAD3-dependent signaling by selectively blocking formation of TGFB-induced SMAD3-SMAD4 complex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EID2
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q6X7S9, Q8N6I1
EID2
KLH conjugated synthetic peptide between 177-208 amino acids from the C-terminal region of human EID2.
400 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot
Unconjugated
EID2
CREBBP/EP300 inhibitor 2; CREBBP/EP300 inhibitory protein 2; Cri2; E1A like inhibitor of differentiation 2; EID-1-like inhibitor of differentiation 2; Eid2; EID-2; EP300 interacting inhibitor of differentiation 2; EP300-interacting inhibitor of differentiation 2
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
163126, 386655
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.