Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EID2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | EID2B |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
EID2B Polyclonal specifically detects EID2B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EID2B | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EID-2B, EID-2-like inhibitor of differentiation 3, EID3, EID-3EID-2-like inhibitor of differentiation-3, EP300 interacting inhibitor of differentiation 2B, EP300-interacting inhibitor of differentiation 2B, FLJ38944 | |
EID2B | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
Q96D98 | |
126272 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QQLAFLEINRQLLFREYLDGSSMIPVRLLRDFEERRRLFVEGCKA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title