Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EID2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564159 Supplier Invitrogen™ Supplier No. PA564159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQLAFLEINR QLLFREYLDG SSMIPVRLLR DFEERRRLFV EGCKA Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 56%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen EID2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EID2B
Gene Accession No. Q96D98
Gene Alias EID2B; EID-2B; EID-2-like inhibitor of differentiation 3; EID-2-like inhibitor of differentiation-3; EID3; EID-3; EP300 interacting inhibitor of differentiation 2B; EP300-interacting inhibitor of differentiation 2B
Gene Symbols EID2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EID2B. Recombinant protein control fragment (Product #RP-103549).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126272
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.