Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EID3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531556 Supplier Invitrogen™ Supplier No. PA531556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tissue-specific component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination and mediates sumoylation of shelterin complex (telosome) components. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EID3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene EID3
Gene Accession No. Q8N140
Gene Alias 1700027M21Rik; E1A-like inhibitor of differentiation 3; EID-1-like inhibitor of differentiation 3; Eid3; EID-3; eid3 {ECO:0000312; EMBL:AAH97404.1}; EP300 interacting inhibitor of differentiation 3; EP300-interacting inhibitor of differentiation 3; non-SMC element 4 homolog B; non-structural maintenance of chromosomes element 4 homolog B; NS4EB; NSE4B; NSMCE4B; testis tissue sperm-binding protein Li 96mP
Gene Symbols EID3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EID3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493861
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.