Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EID3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551354 Supplier Invitrogen™ Supplier No. PA551354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total EID3 protein.

Tissue-specific component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination and mediates sumoylation of shelterin complex (telosome) components. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EID3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene EID3
Gene Accession No. Q3V124, Q8N140
Gene Alias 1700027M21Rik; E1A-like inhibitor of differentiation 3; EID-1-like inhibitor of differentiation 3; Eid3; EID-3; eid3 {ECO:0000312; EMBL:AAH97404.1}; EP300 interacting inhibitor of differentiation 3; EP300-interacting inhibitor of differentiation 3; non-SMC element 4 homolog B; non-structural maintenance of chromosomes element 4 homolog B; NS4EB; NSE4B; NSMCE4B; testis tissue sperm-binding protein Li 96mP
Gene Symbols EID3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the N terminal of human EP300 interacting inhibitor of differentiation 3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493861, 66341
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.