Learn More
Invitrogen™ eIF1 Monoclonal Antibody (2B9)

Description
MA1-077 detects eIF1 in human, mouse, and rat samples and has been successfully used in Western blot and IF/ICC applications.
Specifications
Specifications
| Antigen | eIF1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2B9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide |
| Gene | Eif1 |
| Gene Accession No. | P41567, P48024 |
| Gene Alias | A121; EIF1; EIF-1; eIF13; EIF1A; Eukaryotic translation initiation factor 1; ISO1; Protein translation factor SUI1 homolog; SUI1; sui1iso1; Sui1-rs1; suppressor of initiator codon mutations, related sequence 1; suppressor of initiator codon mutations-Yeast homolog related sequence 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.