Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF1 Mouse anti-Human, Clone: 2B9, Abnova™

Catalog No. 89035443 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-443 Supplier Abnova Corporation Supplier No. MAB14038

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human EIF1.

  • High specificity
  • High reactivity

Specifications

Antigen EIF1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2B9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human EIF1.
Dilution Flow Cytometry (3-5 ug x 106 cells) Immunocytochemistry (1-2 ug/mL) Immunofluorescence (1-2 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P41567
Gene Alias A121/EIF-1/EIF1A/ISO1/SUI1
Gene Symbols EIF1
Host Species Mouse
Immunogen Full length human EIF1 produced in HEK293T cells.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10209
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.