Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588038 Supplier Invitrogen™ Supplier No. PA588038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSAIQNLHSF DPFADASKGD DLLPAGTEDY IHIRIQQRNG RKTLTTVQGI ADDYDKKKLV KAFKKKFACN GTVIEHPEYG EVIQLQGDQR KNICQFLVEI GLAKDDQLKV HGF; Positive Samples: A-549, Mouse liver.

Necessary for scanning and involved in initiation site selection. Promotes the assembly of 48S ribosomal complexes at the authentic initiation codon of a conventional capped mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Eif1
Gene Accession No. P41567, P48024
Gene Alias A121; EIF1; EIF-1; eIF13; EIF1A; Eukaryotic translation initiation factor 1; ISO1; Protein translation factor SUI1 homolog; SUI1; sui1iso1; Sui1-rs1; suppressor of initiator codon mutations, related sequence 1; suppressor of initiator codon mutations-Yeast homolog related sequence 1
Gene Symbols Eif1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human EIF1 (NP_0057921).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10209, 20918
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.