Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143916
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143916 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143916 Supplier Invitrogen™ Supplier No. PA5143916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human HEK293 whole cell, human Jurkat whole cell, human Raji whole cell, human K562 whole cell, rat pancreas tissue, mouse pancreas tissue, mouse NIH/3T3 whole cell. IHC: human placenta tissue. ICC/IF: T-47D cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Necessary for scanning and involved in initiation site selection. Promotes the assembly of 48S ribosomal complexes at the authentic initiation codon of a conventional capped mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Eif1
Gene Accession No. P41567, P48024
Gene Alias A121; EIF1; EIF-1; eIF13; EIF1A; Eukaryotic translation initiation factor 1; ISO1; Protein translation factor SUI1 homolog; SUI1; sui1iso1; Sui1-rs1; suppressor of initiator codon mutations, related sequence 1; suppressor of initiator codon mutations-Yeast homolog related sequence 1
Gene Symbols Eif1
Host Species Rabbit
Immunogen E.coli-derived human EIF1 recombinant protein (Position: S2-D106).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10209, 20918, 287703
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.