Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982589
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2589 Supplier Bioss Supplier No. BS13066R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Necessary for scanning and involved in initiation site selection. Promotes the assembly of 48S ribosomal complexes at the authentic initiation codon of a conventional capped mRNA.

Specifications

Antigen eIF1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Eif1
Gene Accession No. P41567
Gene Alias A121; Eif1; EIF-1; eIF13; EIF1A; Eukaryotic translation initiation factor 1; ISO1; Protein translation factor SUI1 homolog; Sui1; Sui1iso1; Sui1-rs1; suppressor of initiator codon mutations, related sequence 1; suppressor of initiator codon mutations-Yeast homolog related sequence 1
Gene Symbols Eif1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human eIF1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10209
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.