Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF1AX Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117033
Description
Positive Samples: SKOV3, HT-29, MCF7, NCI-H460, Mouse brain, Mouse kidney Immunogen sequence: KNRRRGKNEN ESEKRELVFK EDGQEYAQVI KMLGNGRLEA MCFDGVKRLC HIRGKLRKKV WINTSDIILV GLRDYQDNKA DVILKYNADE ARSLKAYGEL PEHAKINETD TFGPGDDDEI QFDDIGDDDE DIDDI.
This gene encodes an essential eukaryotic translation initiation factor. The protein is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
Specifications
eIF1AX | |
Polyclonal | |
Unconjugated | |
EIF1AX | |
1500010B24Rik; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; EIF1AP1; EIF1AX; Eif1ay; EIF4C; eIF-4C; eukaryotic translation initiation factor 1A X-linked; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1964, 302697, 66235 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.9 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P47813, Q8BMJ3 | |
EIF1AX | |
Recombinant fusion protein containing a sequence corresponding to amino acids 10-144 of human EIF1AX (NP_0014031). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction