Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF1AY Antibody (OTI2G2), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271417G
Description
EIF1A Monoclonal specifically detects EIF1A in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
EIF1A | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EIF1AX | |
Recombinant protein fragment corresponding to amino acids 2-144 of human EIF1AY (NP_004672) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot | |
OTI2G2 | |
Western Blot | |
eIF-1A, EIF1AP1, eIF-4C, EIF4C, eukaryotic translation initiation factor 1A, X chromosome, eukaryotic translation initiation factor 1A, X-chromosomal, eukaryotic translation initiation factor 1A, Y chromosome, eukaryotic translation initiation factor 1A, Y-chromosomal, Eukaryotic translation initiation factor 4C, FLJ45762, Putative eukaryotic translation initiation factor 1A | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
1964 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction