Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF1AY Monoclonal Antibody (OTI2G2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807511
Description
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5′ end of capped RNA.Specifications
| EIF1AY | |
| Monoclonal | |
| Unconjugated | |
| EIF1AY | |
| eIF-4C | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9086 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2G2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14602 | |
| EIF1AY | |
| Human recombit protein fragment corresponding to amino acids 2-144 of human EIF1AY produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction