Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF1AY Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576163
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
Specifications
eIF1AY | |
Polyclonal | |
Unconjugated | |
EIF1AY | |
1500010B24Rik; Ab1-287; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; eIF-1A Y isoform; EIF1AP1; EIF1AX; EIF1AY; EIF4C; eIF-4C; Eukaryotic translation initiation factor 1A; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y chromosome; eukaryotic translation initiation factor 1A, Y-chromosomal; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Liver regeneration-related protein LRRG048; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9086 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O14602 | |
EIF1AY | |
Recombinant fusion protein of human EIF1AY. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction