Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF1AY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116193
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116193 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116193 Supplier Invitrogen™ Supplier No. PA5116193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF1AY Polyclonal Antibody for IHC (P), Western Blot

This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF1AY
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene EIF1AY
Gene Accession No. O14602
Gene Alias 1500010B24Rik; Ab1-287; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; eIF-1A Y isoform; EIF1AP1; EIF1AX; EIF1AY; EIF4C; eIF-4C; Eukaryotic translation initiation factor 1A; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y chromosome; eukaryotic translation initiation factor 1A, Y-chromosomal; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Liver regeneration-related protein LRRG048; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A
Gene Symbols EIF1AY
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human EIF1AY.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9086
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.