Learn More
EIF1AY Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 111931AP150UL

Description
This is a rabbit polyclonal antibody raised against the full length of human EIF1AY, and can recognize both EIF1AY and EIF1AX.
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5′ end of capped RNA.Specifications
EIF1AY | |
Polyclonal | |
Unconjugated | |
EIF1AY | |
eIF 1A Y isoform, eIF 4C, EIF1AY | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1964, 9086 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14602, P47813 | |
EIF1AX, EIF1AY | |
EIF1AY Fusion Protein Ag1680 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.