Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EIF1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568190 Supplier Invitrogen™ Supplier No. PA568190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

EIF1B is probably involved in translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF1B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EIF1B
Gene Accession No. O60739, Q9CXU9
Gene Alias 1500010M16Rik; 3110001N14Rik; Eif1b; Eukaryotic translation initiation factor 1b; Gc20; Protein translation factor SUI1 homolog GC20; RGD1309241; translation factor sui1 homolog
Gene Symbols EIF1B
Host Species Rabbit
Immunogen Synthesized peptide derived from human EIF1B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10289, 301068, 68969
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.