Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF1B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571925
Description
EIF1B is probably involved in translation.
Specifications
EIF1B | |
Polyclonal | |
Unconjugated | |
EIF1B | |
Eukaryotic translation initiation factor 1b; eIF1b; Protein translation factor SUI1 homolog GC20; EIF1B | |
Rabbit | |
Protein A | |
RUO | |
10289 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.48 mg/mL | |
PBS with 0.09% sodium azide | |
O60739 | |
EIF1B | |
KLH conjugated synthetic peptide between 84-113 amino acids from the C-terminal region of human EIF1B | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction