Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF2 alpha/EIF2S1 Antibody (5E10), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197936G
Description
eIF2 alpha/EIF2S1 Monoclonal specifically detects eIF2 alpha/EIF2S1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
eIF2 alpha/EIF2S1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EIF2S1 | |
Recombinant human EIF2S1 (1-315aa) purified from E. coli | |
100 μL | |
Phospho Specific | |
1965 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
50000000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human EIF2S1 mAb, clone 5E10, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human EIF2S1 protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction